Learn More
Cyclin E2 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-11935-100UL
Description
The Cyclin E2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Cyclin E2. This antibody recognizes human antigen. The Cyclin E2 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
| Cyclin E2 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| CoraLite 488 | |
| 100 μL | |
| 9134 | |
| IgG |