missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin E2 Antibody (40-89-R), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-32237
This item is not returnable.
View return policy
Description
Cyclin E2 Monoclonal antibody specifically detects Cyclin E2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Cyclin E2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| CYCE2, cyclin E2, G1/S-specific cyclin E2, G1/S-specific cyclin-E2 | |
| Synthetic peptide within Human Cyclin E2 aa 1-50/ 404. (Uniprot: O96020) | |
| 100 μL | |
| Cancer, Cell Cycle and Replication, Cellular Markers | |
| 9134 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| 40-89-R | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction