missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin E1 Rabbit anti-Human, Clone: CCNE1/4935R, Novus Biologicals™
Rabbit Monoclonal Antibody
£245.00
Specifications
| Antigen | Cyclin E1 |
|---|---|
| Clone | CCNE1/4935R |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
Descrizione
Cyclin E1 Monoclonal specifically detects Cyclin E1 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifica
| Cyclin E1 | |
| 0.2 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Ovarian Carcinoma Cell Markers, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 898 | |
| IgG κ | |
| Protein A or G purified |
| CCNE1/4935R | |
| Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1 | |
| Recombinant human Cyclin E1 protein fragment (around aa 10-176) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto