missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin E1 Mouse anti-Human, Clone: CCNE1/2460, Novus Biologicals™
Mouse Monoclonal Antibody
£218.00
Specifications
| Antigen | Cyclin E1 |
|---|---|
| Clone | CCNE1/2460 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin, Protein Array |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array |
Description
Cyclin E1 Monoclonal specifically detects Cyclin E1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Cyclin E1 | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Ovarian Carcinoma Cell Markers, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 898 | |
| IgG2b κ | |
| Protein A or G purified |
| CCNE1/2460 | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1 | |
| Recombinant human Cyclin E1 protein fragment (around aa 10-176) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title