missing translation for 'onlineSavingsMsg'
Learn More

Cyclin E1 Antibody (3A5), Novus Biologicals™

Rabbit Monoclonal Antibody

£269.00 - £406.00

Specifications

Antigen Cyclin E1
Clone 3A5
Dilution ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200
Applications ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043136
View Documents
Novus Biologicals
NBP3-26692-50ul
50 μL
£269.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043323
View Documents
Novus Biologicals
NBP3-26692-100ul
100 μL
£406.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cyclin E1 Monoclonal antibody specifically detects Cyclin E1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry
Specifications

Specifications

Cyclin E1
ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200
Monoclonal
Purified
RUO
Human
CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1
A synthesized peptide derived from Human Cyclin E1 [UniProt P24864]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
3A5
ELISA, Immunohistochemistry, Immunocytochemistry
Unconjugated
Rabbit
Cancer, Cell Cycle and Replication, Cellular Markers, Ovarian Carcinoma Cell Markers, Phospho Specific
PBS, pH 7.4, 150mM NaCl and 50% glycerol
898
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.