missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D3 Rabbit anti-Human, Mouse, Rat, Clone: 4C5C8, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Cyclin D3 Monoclonal antibody specifically detects Cyclin D3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Cyclin D3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 4C5C8 |
| Conjugate | Unconjugated |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | cyclin D3, D3-type cyclin, G1/S-specific cyclin D3, G1/S-specific cyclin-D3 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 202-292 of human Cyclin D3 (P30281). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?