missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cyclin D3 Antibody (DCS2.2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30977-25ug
This item is not returnable.
View return policy
Description
Cyclin D3 Monoclonal antibody specifically detects Cyclin D3 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| Cyclin D3 | |
| Monoclonal | |
| Unconjugated | |
| cyclin D3, D3-type cyclin, G1/S-specific cyclin D3, G1/S-specific cyclin-D3 | |
| The immunogen for the mouse anti-human Cyclin D3 was a recombinant full length (34 kDa) Cyclin D3 protein. | |
| 25 μg | |
| Cancer, Cell Cycle and Replication, Wnt Signaling Pathway | |
| 896 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| DCS2.2 | |
| Phosphate buffered saline | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction