missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin D2 Recombinant Rabbit Monoclonal Antibody (5B4M7)

Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | Cyclin D2 |
|---|---|
| Clone | 5B4M7 |
| Concentration | 0.63 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30283449
|
Invitrogen™
MA558015 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30284714
![]() |
Invitrogen™
MA558016 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: LLGAVCMFL ASKLKETSPL TAEKLCIYTD NSIKPQELLE WELVVLGKLK WNLAAVTPHD FIEHILRKLP QQREKLSLIR KHAQTFIALC ATDFKFAMYP PS.
Cyclin D2 is a G1 cyclin required for G1 phase progression and is a strong candidate for a proto-oncogene. Cyclin D2 can phosphorylate pRB when associated with cdk 4 and / or cdk 6.Specifications
| Cyclin D2 | |
| 0.63 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P30279 | |
| 894 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Cyclin D2 (NP_001750.1). | |
| Antibody |
| 5B4M7 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| CCND2 | |
| 2600016F06Rik; AI256817; BF642806; C86853; CCND2; cD2; cyclin D2; Cyl-2; G1/S-specific cyclin-D2; KIAK0002; MGC102758; MPPH3; Vin1; Vin-1; Vin-1 proto-oncogene | |
| CCND2 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title