missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cyclin D2 Antibody (DCS3.1), Novus Biologicals™
Mouse Monoclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | Cyclin D2 |
|---|---|
| Clone | DCS3.1 |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067164
|
Novus Biologicals™
NBP3-30976-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067110
|
Novus Biologicals™
NBP3-30976-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin D2 Monoclonal antibody specifically detects Cyclin D2 in Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| Cyclin D2 | |
| Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Wnt Signaling Pathway | |
| Phosphate buffered saline | |
| 894 | |
| IgG2a | |
| Ammonium sulfate precipitation |
| DCS3.1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| cyclin D2, G1/S-specific cyclin D2, G1/S-specific cyclin-D2, KIAK0002, MGC102758 | |
| The immunogen for the production of this mouse monoclonal anti-Cyclin D2 was a his-tagged recombinant full length Cyclin D2 34 kDa protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title