missing translation for 'onlineSavingsMsg'
Learn More

Cyclin D1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£159.65 - £373.33

Specifications

Antigen Cyclin D1
Concentration 1000 μg/mL
Applications Flow Cytometry (Intracellular Staining)
Conjugate CoraLite Plus 488
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18075265
View Documents
Proteintech
CL488-26939-20UL
20 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18007065
View Documents
Proteintech
CL488-26939-100UL
100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

The Cyclin D1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Cyclin D1. This antibody recognizes human, mouse, rat antigen. The Cyclin D1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.

Specifications

Specifications

Cyclin D1
Flow Cytometry (Intracellular Staining)
Liquid
Human, Mouse, Rat
B cell lymphoma 1 protein, CCND1, cyclin D1, D11S287E, G1/S specific cyclin D1, PRAD1, PRAD1 oncogene, U21B31
Primary
1000 μg/mL
CoraLite Plus 488
Rabbit
595
IgG
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.