missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D1 Mouse, Clone: SPM587, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
£242.00 - £402.00
Specifications
| Antigen | Cyclin D1 |
|---|---|
| Clone | SPM587 |
| Concentration | 0.2mg/mL |
| Dilution | Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1 - 2ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18138992
|
Novus Biologicals
NBP2-32840-0.02MG |
0.02 mg |
£242.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18062944
|
Novus Biologicals
NBP2-32840-0.1MG |
0.1 mg |
£402.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin D1 Monoclonal specifically detects Cyclin D1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Cyclin D1 | |
| 0.2mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31 | |
| CCND1 | |
| IgG2a κ | |
| Protein A purified | |
| Recognizes a protein of 36kDa, identified as cyclin D1. Cyclin D1, one of the key cell cycle regulators, is a putative proto-oncogene overexpressed in a wide variety of human neoplasms. This antibody neutralizes the activity of cyclin D1 in vivo. About 60% of mantle cell lymphomas (MCL) contain a t(11; 14)(q13; q32) translocation resulting in over-expression of cyclin D1. This antibody is useful in identifying mantle cell lymphomas (cyclin D1 positive) from CLL/SLL and follicular lymphomas (cyclin D1 negative). Occasionally, hairy cell leukemia and plasma cell myeloma weakly express Cyclin D1. |
| SPM587 | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1 - 2ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Primate | |
| P24385, P24385, P25322, P39948 | |
| 595 | |
| Human recombinant full length cyclin D1 protein | |
| Primary | |
| Store at 4C. | |
| 36 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title