missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100706
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Cyclin C.
Cyclin C was originally identified by a genetic screen for human and Drosophila cDNAs that complement a triple knock out of the CLN genes in Saccharomyces cerevisiae. Cyclin C is a nuclear protein present in a multiprotein complex. It interacts both in vitro and in vivo with cdk 8, a novel protein kinase of the cdk family. It is suggested that cyclin C and cdk 8 control RNA polymerase II function. Human cdk 8 cyclin C might be functionally associated with the mammalian transcription apparatus, perhaps involved in relaying growth regulatory signals.
Specifications
| Cyclin C | |
| Polyclonal | |
| Unconjugated | |
| CCNC | |
| AI451004; AU020987; CCNC; CG1C; CycC; cyclin C; cyclin-C; dJ199J3.4 (cyclin C); hSRB11; SRB11 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114839, 51813, 892 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P24863, P39947, Q62447 | |
| CCNC | |
| A synthesized peptide derived from human CCNC(Accession P24863), corresponding to amino acid residues E249-S283. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction