missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21644-1-AP-150UL
This item is not returnable.
View return policy
Description
Cyclin B2 is a member of the cyclin family, specifically the B-type cyclins. The B-type cyclins, B1 and B2, associate with p34cdc2 and are essential components of the cell cycle regulatory machinery. B1 and B2 differ in their subcellular localization. Cyclin B1 co-localizes with microtubules, whereas cyclin B2 is primarily associated with the Golgi region. Cyclin B2 also binds to transforming growth factor beta RII and thus cyclin B2/cdc2 may play a key role in transforming growth factor beta-mediated cell cycle control.Specifications
| Cyclin B2 | |
| Polyclonal | |
| Unconjugated | |
| CCNB2 | |
| CCNB2, cyclin B2, G2/mitotic specific cyclin B2, HsT17299 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12442, 363088, 9133 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95067, P30276 | |
| Ccnb2 | |
| Cyclin B2 Fusion Protein Ag16329 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction