missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin B2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580415
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Cyclin B2.
Cyclin B2 is a member of the cyclin family, specifically the B-type cyclins. The B-type cyclins, B1 and B2, associate with p34cdc2 and are essential components of the cell cycle regulatory machinery. B1 and B2 differ in their subcellular localization. Cyclin B1 co-localizes with microtubules, whereas cyclin B2 is primarily associated with the Golgi region. Cyclin B2 also binds to transforming growth factor beta RII and thus cyclin B2/cdc2 may play a key role in transforming growth factor beta-mediated cell cycle control.
Specifications
| Cyclin B2 | |
| Polyclonal | |
| Unconjugated | |
| Ccnb2 | |
| C-B2; CCNB2; ccnb2.S; Cycb2; cyclin B2; cyclin B2 S homeolog; cyclinb2; G2/mitotic-specific cyclin B2; G2/mitotic-specific cyclin-B2; HsT17299; XELAEV_18020679mg | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 9133 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with no preservative | |
| O95067 | |
| Ccnb2 | |
| A synthetic peptide corresponding to the N-terminus of the Human cyclin B2/CCNB2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction