missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B2 Mouse anti-Human, Mouse, Rat, Xenopus, Clone: X29.2, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | Cyclin B2 |
|---|---|
| Clone | X29.2 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA |
| Applications | ELISA |
Description
Cyclin B2 Monoclonal specifically detects Cyclin B2 in Human, Mouse, Rat, Xenopus samples. It is validated for ELISA.Specifications
| Cyclin B2 | |
| 0.2 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 9133 | |
| IgG1 κ | |
| Protein A or G purified |
| X29.2 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Xenopus species | |
| cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299 | |
| Recombinant full-length Xenopus laevis Cyclin B2 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title