Learn More
Cyclin B2 Mouse anti-Human, CoraLite 488, Clone: 2G8B7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-67726-100UL
Product Code. 17264756
  • £375.00 / 100µL
Description
Cyclin B2 (CCNB2) is a member of cyclin family proteins, which regulate the activities of cyclin dependent kinases (CDKs) and different cyclins function spatially and temporally in specific phases of the cell cycle. Cyclin B2 serves a key role in progression of G2/M transition. Cyclin B2 has been found to be up-regulated in a variety of human cancers, such as adrenocortical carcinoma, breast carcinoma, colorectal adenocarcinoma, pituitary adenoma and gastric cancer. The aberrant expression of Cyclin B2 deregulates spindle checkpoints in the cell cycle and results in chromosomal instability (CIN), one of the signature phenotypes of most cancers. Moreover, serum circulating Cyclin B2 mRNA expression has been found increased in cancer patients and associated with cancer stage and metastasis status.
Specifications
Specifications
Cyclin B2
Monoclonal
1000 μg/mL
Mouse
Primary
Human
IgG2a
Immunofluorescence
2G8B7
CoraLite 488
100 μL
9133