missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cyclin B2 Monoclonal specifically detects Cyclin B2 in Human, Mouse, Rat, Xenopus samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Cyclin B2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | X29.2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 10mM PBS |
| Gene Alias | cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299 |
| Host Species | Mouse |
| Immunogen | Recombinant full-length Xenopus laevis Cyclin B2 protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?