Learn More
Cyclin B1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-28603-100UL
Description
The Cyclin B1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Cyclin B1. This antibody recognizes human antigen. The Cyclin B1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| Cyclin B1 | |
| 1000 μg/mL | |
| CCNB, CCNB1, cyclin B1, G2/mitotic specific cyclin B1 | |
| 100 μL | |
| 891 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |