missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B1 Antibody (PSA1.1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50189
This item is not returnable.
View return policy
Description
Cyclin B1 Monoclonal antibody specifically detects Cyclin B1 in Human samples. It is validated for Western Blot
Specifications
| Cyclin B1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000 | |
| CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1 | |
| PSATGKVIDKKLPC | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Mitotic Regulators, Tumor Suppressors | |
| 891 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgM |
| Western Blot | |
| PSA1.1 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Thiophilic Chromatography | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction