missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£410.00
Specifications
| Antigen | Cyclin B1 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Cyclin B1 Polyclonal specifically detects Cyclin B1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
| Cyclin B1 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Biology, Cell Cycle and Replication, Mitotic Regulators, Tumor Suppressors | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 891 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PVPVSEPVPEPEPEPEPEPVKEEKLSPEPILVDTASPSPMETSGCAPAEEDLCQAFSDVILAVNDVDAEDGAD | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
| Unconjugated | |
| RUO | |
| Human | |
| CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1 | |
| CCNB1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title