missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B1 Antibody (5G6), Novus Biologicals™
Mouse Monoclonal Antibody
£161.00
Specifications
| Antigen | Cyclin B1 |
|---|---|
| Clone | 5G6 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Cyclin B1 Monoclonal specifically detects Cyclin B1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Cyclin B1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 891 | |
| Purified recombinant fragment of human CCNB1 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 5G6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cell Cycle and Replication, Mitotic Regulators, Tumor Suppressors | |
| CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1 | |
| CCNB1 | |
| IgG1 | |
| Unpurified | |
| 60 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title