missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5119988
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Positive Control: Hela, MCF-7, 293, F9, MEF-1 Subcellular Location: Early/ Late endosome
Cyclin A2 Polyclonal antibody specifically detects Cyclin A2 in Human, Mouse, Rat samples. It is validated for Immunoprecipitation, Western Blot
Tekniske data
| Cyclin A2 | |
| Polyclonal | |
| Unconjugated | |
| CCNA2 | |
| AA408589; Ccn1; Ccn-1; CCNA; CCNA2; ccna2.S; Cyca; CycA2; Cyclin A; cyclin A2; Cyclin A2 (Cyclin A); cyclin A2 S homeolog; Cyclin A-3; cyclin-A; Cyclin-A2; XELAEV_18009326mg | |
| Synthetic peptide within C-terminal human Cyclin A2 | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P20248, P51943 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114494, 12428, 890 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion