missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin A2 Mouse anti-Human, Clone: CCNA2/2333, Novus Biologicals™
Mouse Monoclonal Antibody
£405.00
Specifications
| Antigen | Cyclin A2 |
|---|---|
| Clone | CCNA2/2333 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
Cyclin A2 Monoclonal specifically detects Cyclin A2 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Cyclin A2 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors | |
| 10 mM PBS with 0.05% BSA | |
| 890 | |
| IgG2a κ | |
| Protein A or G purified |
| CCNA2/2333 | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CCN1CCNAcyclin-A, cyclin A2, Cyclin-A, cyclin-A2 | |
| Full length recombinant Cyclin A2 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title