missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin A2 Monoclonal Antibody (N0-E3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA544604
This item is not returnable.
View return policy
Description
Cyclin A2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Cyclins are regulatory subunits of the cyclin-dependent kinases (cdks) and they control transition at different specific phases of the cell cycle. The temporal expression of cyclins is tightly regulated and subsequently plays a critical role in controlling the enzymatic activity of cdks. These cyclin / cdk complexes are essential for passage through specific stages in the cell cycle. In mammalian somatic cells, cyclin A is required for S phase and passage through G2-phase.
Specifications
| Cyclin A2 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P20248, P51943 | |
| CCNA2 | |
| Recombinant protein within human p53 aa 50-432. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| N0-E3 | |
| Unconjugated | |
| CCNA2 | |
| AA408589; Ccn1; Ccn-1; CCNA; CCNA2; ccna2.S; Cyca; CycA2; Cyclin A; cyclin A2; Cyclin A2 (Cyclin A); cyclin A2 S homeolog; Cyclin A-3; cyclin-A; Cyclin-A2; XELAEV_18009326mg | |
| Mouse | |
| Protein A | |
| RUO | |
| 114494, 12428, 890 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu