missing translation for 'onlineSavingsMsg'
Learn More

Cyclin A2 Antibody (E67) - Azide and BSA Free, Novus Biologicals™

Product Code. 18420612 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420612 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420612 Supplier Novus Biologicals Supplier No. NBP2346130.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cyclin A2 Monoclonal antibody specifically detects Cyclin A2 in Human, Mouse, Bovine, Mink, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin A2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone E67
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1 μg/mL, Flow Cytometry 0.5-1 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, CyTOF-ready
Formulation 10 mM PBS
Gene Alias CCN1CCNAcyclin-A, cyclin A2, Cyclin-A, cyclin-A2
Host Species Mouse
Immunogen Full length bovine Cyclin A2 protein (Uniprot: P20248)
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 890
Target Species Human, Mouse, Bovine, Mink, Monkey
Content And Storage Store at -20°C to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.