missing translation for 'onlineSavingsMsg'
Learn More

Cyclin A1 Mouse anti-Xenopus, Clone: XLA1-1, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-07829-100ug

 View more versions of this product

Product Code. 18653649

  • £377.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cyclin A1 Monoclonal specifically detects Cyclin A1 in Xenopus samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cyclin A1
Monoclonal
0.2 mg/mL
Western Blot 1 to 2 μg/mL
cyclin A1, cyclin-A1
Recombinant full-length Xenopus laevis Cyclin A1 protein
100 μg
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors
8900
Store at 4°C.
IgG1 κ
Western Blot
XLA1-1
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Primary
Xenopus species
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.