missing translation for 'onlineSavingsMsg'
Learn More

Cyclin A1/Cyclin A2 Rabbit anti-Human, Mouse, Clone: 10O1C8, Novus Biologicals™

Product Code. 18312322
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18312322 20 μg 20µL
18332932 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18312322 Supplier Novus Biologicals Supplier No. NBP31614020UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cyclin A1/Cyclin A2 Monoclonal antibody specifically detects Cyclin A1/Cyclin A2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin A1/Cyclin A2
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 10O1C8
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 250-465 of human Cyclin A1/Cyclin A2 (P20248).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.