missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin A1/Cyclin A2 Rabbit anti-Human, Clone: S07-3D4, Novus Biologicals™
Rabbit Monoclonal Antibody
£222.00 - £451.00
Specifications
| Antigen | Cyclin A1/Cyclin A2 |
|---|---|
| Clone | S07-3D4 |
| Dilution | Western Blot |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18381597
|
Novus Biologicals
NBP3-14867-25UL |
25 μg |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18331145
|
Novus Biologicals
NBP3-14867-100UL |
100 μg |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin A1/Cyclin A2 Monoclonal antibody specifically detects Cyclin A1/Cyclin A2 in Human samples. It is validated for Western BlotSpecifications
| Cyclin A1/Cyclin A2 | |
| Western Blot | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| IgG | |
| Affinity purified | |
| Antibody |
| S07-3D4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| A synthesized peptide derived from human Cyclin A1/Cyclin A2 (Uniprot#: P20248 P78396) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title