missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cyclin A1 Monoclonal antibody specifically detects Cyclin A1 in Xenopus samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Cyclin A1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | XLA1-1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100 - 1:2000 |
| Formulation | PBS |
| Gene Alias | cyclin A1, cyclin-A1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?