missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin A1/A2 Recombinant Rabbit Monoclonal Antibody (23GB3525), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Cyclin A1/A2 |
|---|---|
| Clone | 23GB3525 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088222
![]() |
Invitrogen™
MA553376 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088206
|
Invitrogen™
MA553377 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin A1/A2 Recombinant Monoclonal antibody specifically detects Cyclin A1/A2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Cyclin A1/A2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P20248, P51943, P78396, Q61456, Q6AY13 | |
| 114494, 12427, 12428, 295052, 890, 8900 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3525 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AA408589; CCN1; Ccn-1; CCNA; Ccna1; CCNA2; CT146; Cyca; Cyca2; Cyclin A; cyclin A1; cyclin A2; Cyclin A2 (Cyclin A); CyclinA; cyclin-A; CyclinA1; Cyclin-A1; cyclin-A2; testicular tissue protein Li 34 | |
| A synthetic peptide of human Cyclin A1/A2 (250-465AA). | |
| Primary | |
| -20°C | |
| CCNA2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title