missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536048
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 52 kDa. Purity is >95% by SDS-PAGE.
The critical role that the family of regulatory proteins known as cyclins play in eukaryotic cell cycle regulation is well established. The best-characterized cyclin complex is the mitotic cyclin B/Cdc2 p34 kinase, the active component of maturing promoting factor. Cyclin A accumulates prior to cyclin B in the cell cycle, appears to be involved in control of S phase and has been shown to associate with cyclin-dependent kinase-2 (Cdk2). In addition, cyclin A has been implicated in cell transformation and is found in complexes with E1A, transcription factors DRTF1 and E2F and retinoblastoma protein, p110. A second form of cyclin A, named cyclin A1 because of its high sequence homology to Xenopus cyclin A1, is most highly expressed in germ cells. It has been proposed that cyclin A1 can associate with Cdk2, p39 and Cdc2 p34.
Specifications
| Cyclin A | |
| Polyclonal | |
| Unconjugated | |
| CCNA2 | |
| AA408589; CCN1; Ccn-1; CCNA; Ccna1; CCNA2; CT146; Cyca; Cyca2; Cyclin A; cyclin A1; cyclin A2; Cyclin A2 (Cyclin A); CyclinA; cyclin-A; CyclinA1; Cyclin-A1; cyclin-A2; testicular tissue protein Li 34 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114494, 12427, 12428, 295052, 890, 8900 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P20248, P51943, P78396, Q61456, Q6AY13 | |
| CCNA1, CCNA2 | |
| Synthetic peptide corresponding to amino acids 220-270 of human Cyclin A. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction