missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Cyclic Pifithrin-Alpha (Hydrobromide)
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
- Cyclic Pifthrin-Alpha is a cell-permeable and reversible inhibitor of p53-mediated apoptosis and p53-dependent gene transcription
- It is a more stable and less cytotoxic analog of the non-cyclic form of pifithrin-alpha
- Cyclic Pifithrin-Alpha has also been reported to activate the aryl hydrocarbon receptor
- (Fernandez-Cruz et al.), Gary & Jensen, Komarov et al.)
- Increases efficiency of reprogramming mouse embryonic fibroblasts to induced pluripotent stem cells (Liao et al.)
- Reduces UV-induced apoptosis of mouse embryonic stem cells (Qin et al.)
- Increases the numbers of mouse hematopoietic stem and progenitor cells in vivo and in vitro, also decreases the radiation-induced death of these cells (Leonova et al.)
Specifications
Specifications
| CAS | 511296-88-1 |
| Synonym | Cyclic PFT-α; Cyclic Pifithrin-α; PFT-β; Pifithrin-β |
| Quantity | 5 mg |
| Molecular Formula | C16H16N2S · HBr |
| Purity | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction