missing translation for 'onlineSavingsMsg'
Learn More

Cyclic AMP, MedChemExpress

Product Code. 16442349
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442349 1 g 1g
16402286 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16442349 Supplier Medchem Express Supplier No. HYB1511
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cyclic AMP (cAMP) is a mitogenic messenger and promotes the G1 to S phase transition in the cell cycle.

Cyclic AMP (cAMP) is a mitogenic messenger and promotes the G1 to S phase transition in the cell cycle.

Specifications

Chemical Name or Material Cyclic AMP
CAS 60-92-4
Color White
Purity Grade Notes Research
Molecular Formula C10H12N5O6P
Quantity 1 g
Synonym Cyclic adenosine monophosphate Adenosine cyclic 3', 5'-monophosphate cAMP
Solubility Information 0.1 M NaOH : 30 mg/mL (91.13 mM; ultrasonic and adjust pH to 6 with NaOH) ∣H2O : 2.4 mg/mL (7.29 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (3.04 mM; ultrasonic and warming and heat to 60°C)
SMILES NC1=C2N=CN([C@@H]3O[C@]4([H])COP(O)(O[C@@]4([H])[C@H]3O)=O)C2=NC=N1
Molecular Weight (g/mol) 329.21
Formula Weight 329.21
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.