missing translation for 'onlineSavingsMsg'
Learn More

Cyclic 17-(1,2-Ethanediyl Acetal) Androst-4-ene-3,17-dione, TRC

Product Code. 30364533
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364533 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30364533 Supplier Toronto Research Chemicals Supplier No. TRCC990020250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclic 17-(1,2-Ethanediyl Acetal) Androst-4-ene-3,17-dione
CAS 1044-89-9
Molecular Formula C21H30O3
Quantity 250 mg
Synonym 17,17-(Ethylenedioxy)androst-4-en-3-one
InChI Formula InChI=1S/C21H30O3/c1-19-8-5-15(22)13-14(19)3-4-16-17(19)6-9-20(2)18(16)7-10-21(20)23-11-12-24-21/h13,16-18H,3-12H2,1-2H3/t16-,17+,18+,19+,20+/m1/s1
SMILES C[C@@]12C3(CC[C@]1([C@]4([C@](CC2)([C@]5(C)C(CC4)=CC(=O)CC5)[H])[H])[H])OCCO3
IUPAC Name (8R,9S,10R,13S,14S)-10,13-dimethyl-1,6,7,8,9,10,11,12,13,14,15,16-dodecahydrospiro[cyclopenta[a]phenanthrene-17,2'-[1,3]dioxolan]-3(2H)-one
Molecular Weight (g/mol) 330.46
Formula Weight 330.22
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.