missing translation for 'onlineSavingsMsg'
Learn More

Cyclen, TRC

Product Code. 30134716
Change view
Click to view available options
Quantity:
250 mg
5 g
25 g
Unit Size:
250mg
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134716 25 g 25g
30133036 5 g 5g
30007513 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134716 Supplier Toronto Research Chemicals Supplier No. TRCC98774525G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 294-90-6
Molecular Formula C8 H20 N4
Molecular Weight (g/mol) 172.27
Synonym Gadobutrol Impurity 39 (Cyclen)
IUPAC Name 1,4,7,10-tetrazacyclododecane
SMILES C1CNCCNCCNCCN1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H20N4/c1-2-10-5-6-12-8-7-11-4-3-9-1/h9-12H,1-8H2
Formula Weight 172.1688
Chemical Name or Material Cyclen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.