missing translation for 'onlineSavingsMsg'
Learn More

Cyclen, 97%

Product Code. 10348780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10348780 250 mg Glass bottle 250mg
10687114 50 mg Glass bottle 50mg
10036153 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10348780 Supplier Thermo Scientific Acros Supplier No. 294322500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 294-90-6 | C8H20N4 | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 294-90-6
Molecular Formula C8H20N4
Molecular Weight (g/mol) 172.27
MDL Number MFCD00066281
InChI Key QBPPRVHXOZRESW-UHFFFAOYSA-N
Synonym cyclen, 1,4,7,10-tetraazacyclododecane, 12 anen4, 1,4,7,10tetraaza-cyclododecane, 5-26-11-00023 beilstein handbook reference, 1,4,7,10-tetracyclododecane, 1,4,7,10-tetraazacyclododecane cyclen, tetraaza-12-crown-4, pubchem23801
PubChem CID 64963
ChEBI CHEBI:37391
IUPAC Name 1,4,7,10-tetrazacyclododecane
SMILES C1CNCCNCCNCCN1

Specifications

Melting Point 108.0°C to 113.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 2699
Solubility Information Solubility in water: soluble
Formula Weight 172.27
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Cyclen, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclen
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.