missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclamic acid, MedChemExpress
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
missing translation for 'unitSize'
1g
5g
Description
Cyclamic acid (Cyclohexylsulfamic acid) is one of the most widely used artificial sweetenersin food and pharmaceuticals.
Specifications
Specifications
| Chemical Name or Material | Cyclamic acid |
| Physical Form | Solid |
| CAS | 100-88-9 |
| Color | White |
| Quantity | 5 g |
| Purity Grade Notes | Research |
| Molecular Formula | C6H13NO3S |
| Synonym | Cyclohexylsulfamic acid Cyclamate |
| Solubility Information | DMSO : 100 mg/mL (557.91 mM; Need ultrasonic) |
| SMILES | O=S(O)(NC1CCCCC1)=O |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction