missing translation for 'onlineSavingsMsg'
Learn More

CYC-116, MedChemExpress

Product Code. 16460397
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16460397

Brand: Medchem Express HY10558

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CYC-116 is a potent aurora A and aurora B inhibitor with Kis of 8 and 9 nM, respectively.

CYC-116 is a potent aurora A and aurora B inhibitor with Kis of 8 and 9 nM, respectively.

Specifications

Chemical Name or Material CYC-116
Physical Form Solid
CAS 693228-63-6
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H20N6OS
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 15 mg/mL (40.71 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC1=C(SC(N)=N1)C2=NC(NC3=CC=C(C=C3)N4CCOCC4)=NC=C2
Molecular Weight (g/mol) 368.46
Formula Weight 368.46
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.