missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYBRD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£139.05 - £359.47
Specifications
| Antigen | CYBRD1 |
|---|---|
| Concentration | 0.63 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16825115
|
Proteintech
26735-1-AP-20UL |
20 μL |
£139.05
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16815115
|
Proteintech
26735-1-AP-150UL |
150 μL |
£359.47
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene is a member of the cytochrome b(561) family that encodes an iron-regulated protein. It highly expressed in the duodenal brush border membrane. It has ferric reductase activity and is believed to play a physiological role in dietary iron absorption.Specifications
| CYBRD1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q53TN4 | |
| 79901 | |
| CYBRD1 Fusion Protein Ag24968 | |
| Primary | |
| -20°C | |
| CYBRD1 |
| 0.63 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CYBRD1, cytochrome b reductase 1, DCYTB, Duodenal cytochrome b, Ferric chelate reductase 3, FRRS3 | |
| CYBRD1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title