missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYBRD1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142130
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognise isoform 1 (NP_079119.3) only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
This gene is a member of the cytochrome b(561) family that encodes an iron-regulated protein. It highly expressed in the duodenal brush border membrane. It has ferric reductase activity and is believed to play a physiological role in dietary iron absorption.
Specifications
| CYBRD1 | |
| Polyclonal | |
| Unconjugated | |
| CYBRD1 | |
| 2210407P13Rik; CYB561A2; CYBRD1; Cytochrome b reductase 1; cytochrome b561 family, member A2; Dcytb; Duodenal cytochrome b; ferric-chelate reductase 3; FRRS3 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 79901 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q53TN4 | |
| CYBRD1 | |
| Peptide with sequence CRNLALDEAGQRSTM. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction