missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYBB/NOX2 Antibody (NL7), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-41012PCP
This item is not returnable.
View return policy
Description
CYBB/NOX2 Monoclonal specifically detects CYBB/NOX2 in Human samples. It is validated for Flow Cytometry, Flow (Intracellular).
Specifications
| CYBB/NOX2 | |
| Monoclonal | |
| PerCP | |
| P04839 | |
| CYBB | |
| Cytochrome B245 solubilised in octyl-beta-glucopyranoside | |
| 0.1 mL | |
| Primary | |
| This antibody recognises the heavy chain of cytochrome B245. It recognises the native as well as the denatured form of the protein | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| NL7 | |
| Flow Cytometry, Flow (Intracellular) | |
| CGD, CGD91-phox, Cytochrome b(558) subunit beta, cytochrome b-245 heavy chain, cytochrome b-245, beta polypeptide, Cytochrome b558 subunit beta, EC 1.6.3, GP91-1, GP91PHOX, GP91-PHOX, Heme-binding membrane glycoprotein gp91phox, NADPH oxidase 2, Neutrophil cytochrome b 91 kDa polypeptide, NOX2chronic granulomatous disease, p22 phagocyte B-cytochrome, p91-PHOX, Superoxide-generating NADPH oxidase heavy chain subunit | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 1536 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction