missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYB5RL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | CYB5RL |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
CYB5RL Polyclonal specifically detects CYB5RL in Human samples. It is validated for Western Blot.Specifications
| CYB5RL | |
| Polyclonal | |
| Rabbit | |
| NP_001026842 | |
| 606495 | |
| Synthetic peptide directed towards the N terminal of human LOC606495. Peptide sequence KLNPETFVAFCIIAMDRLTKDTYRVRFALPGNSQLGLRPGQHLILRGIVD. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| cytochrome b5 reductase-like | |
| CYB5RL | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title