missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYB5R4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103468
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CYB5R4.
NCB5OR is a flavohemoprotein that contains functional domains found in both cytochrome b5 (CYB5A; MIM 250790) and CYB5 reductase (see MIM 250800).
Specifications
| CYB5R4 | |
| Polyclonal | |
| Unconjugated | |
| Cyb5r4 | |
| 2810034J18Rik; b5&b5R; b5/b5r; B5+B5R; b5b5r; C79736; cb5/cb5R; Cb5cb5r; Cyb5r4; cytochrome b5 reductase 4; cytochrome b-type NAD(P)H oxidoreductase; dJ676J13.1; flavohemoprotein b5/b5R; flavohemoprotein b5+b5R; hypothetical protein LOC553685; im:4727699; NADPH cytochrome B5 oxidoreductase; Ncb5or; N-terminal cytochrome b5 and cytochrome b5 oxidoreductase domain-containing protein; RP4-676J13.1; Unknown (protein for MGC:134229); zgc:112177 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51167 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q7L1T6 | |
| Cyb5r4 | |
| A synthesized peptide derived from human CYB5R4(Accession Q7L1T6), corresponding to amino acid residues K64-W114. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction