missing translation for 'onlineSavingsMsg'
Learn More

CYB5R3 Rabbit anti-Human, Mouse, Pig, Rat, CoraLite 594, Polyclonal, Proteintech

Product Code. 17619926 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17619926 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17619926 Supplier Proteintech Supplier No. CL59410894100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CYB5R3 Polyclonal antibody apecifically detetects CYB5R3 antigen in Human, Mouse, Pig, Rat Samples. It is validated for Immunofluorescence.

Specifications

Antigen CYB5R3
Applications Immunofluorescence
Classification Polyclonal
Concentration 1000 μg/mL
Conjugate CoraLite 594
Formulation PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Gene Accession No. BC004821
Gene Symbols CYB5R3
Host Species Rabbit
Immunogen CYB5R3 fusion protein Ag1339
Purification Method Antigen affinity purification
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 1727
Target Species Human, Mouse, Pig, Rat
Content And Storage Store at -20°C. Avoid exposure to light.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.