missing translation for 'onlineSavingsMsg'
Learn More

CYB5R3 Mouse anti-Human, Clone: 9C4D10, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66022-1-IG-20UL

 View more versions of this product

Product Code. 16877525

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CYB5R3 (NADH-cytochrome b5, reductase 3, diaphorase 1) is a 301 amino acid protein encoded by the human gene CYB5R3. CYB5R3 belongs to the flavoprotein pyridine nucleotide cytochrome reductase family and has two naturally occuring isoforms. Isoform 1 is anchored to the cytoplasmic side of the endoplasmic reticulum membrane and mitochondrion outer membrane,while isoform 2 is the soluble form found in erythrocytes. CYB5R3 is involved in the desaturation and elongation of fatty acids, cholesterol biosynthesis,drug metabolism and, in erythrocytes, methemoglobin reduction.
Specifications

Specifications

CYB5R3
Monoclonal
0.89 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P00387
CYB5R3
CYB5R3 Fusion Protein Ag18096
20 μL
Primary
Human
Antibody
IgG2b
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
9C4D10
Unconjugated
CYB5R3
B5R, CYB5R3, Cytochrome b5 reductase, cytochrome b5 reductase 3, DIA1, Diaphorase 1, NADH cytochrome b5 reductase 3
Mouse
Protein A
RUO
1727
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.