missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYB5R3 Mouse anti-Human, Clone: 9C4D10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66022-1-IG-150UL
This item is not returnable.
View return policy
Description
CYB5R3 (NADH-cytochrome b5, reductase 3, diaphorase 1) is a 301 amino acid protein encoded by the human gene CYB5R3. CYB5R3 belongs to the flavoprotein pyridine nucleotide cytochrome reductase family and has two naturally occuring isoforms. Isoform 1 is anchored to the cytoplasmic side of the endoplasmic reticulum membrane and mitochondrion outer membrane,while isoform 2 is the soluble form found in erythrocytes. CYB5R3 is involved in the desaturation and elongation of fatty acids, cholesterol biosynthesis,drug metabolism and, in erythrocytes, methemoglobin reduction.Specifications
| CYB5R3 | |
| Monoclonal | |
| 0.89 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P00387 | |
| CYB5R3 | |
| CYB5R3 Fusion Protein Ag18096 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 9C4D10 | |
| Unconjugated | |
| CYB5R3 | |
| B5R, CYB5R3, Cytochrome b5 reductase, cytochrome b5 reductase 3, DIA1, Diaphorase 1, NADH cytochrome b5 reductase 3 | |
| Mouse | |
| Protein A | |
| RUO | |
| 1727 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction