missing translation for 'onlineSavingsMsg'
Learn More

CYB5R3 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18368083 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368083 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368083 Supplier Novus Biologicals Supplier No. NBP306497AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CYB5R3 Polyclonal antibody specifically detects CYB5R3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB5R3
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias B5RNADH-cytochrome b5 reductase 3, Cytochrome b5 reductase, cytochrome b5 reductase 3, DIA1NADH-cytochrome b5 reductase 3 membrane-bound form, diaphorase (NADH) (cytochrome b-5 reductase), Diaphorase-1, EC 1.6.2.2, NADH-cytochrome b5 reductase 3 soluble form
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CYB5R3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1727
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.