missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYB5R3 Antibody (8H0), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42953
This item is not returnable.
View return policy
Description
CYB5R3 Monoclonal antibody specifically detects CYB5R3 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| CYB5R3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 8H0 | |
| Western Blot 1:1000 - 1:20000, ELISA, Immunohistochemistry 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:500 | |
| B5RNADH-cytochrome b5 reductase 3, Cytochrome b5 reductase, cytochrome b5 reductase 3, DIA1NADH-cytochrome b5 reductase 3 membrane-bound form, diaphorase (NADH) (cytochrome b-5 reductase), Diaphorase-1, EC 1.6.2.2, NADH-cytochrome b5 reductase 3 soluble form | |
| cytochrome b5 reductase 3 - (Uniprot# P00387) | |
| 100 μg | |
| Cancer, Lipid and Metabolism | |
| 1727 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction