missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYB5R2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113008
This item is not returnable.
View return policy
Description
CYB5R2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
CYB5R2 is an NADH-cytochrome b5 reductase which are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction. Responsible for NADH-dependent lucigenin chemiluminescence in spermatozoa by reducing both lucigenin and 2-[4-iodophenyl]-3-[4-nitrophenyl]-5-[2,4-disulfophenyl]-2H tetrazolium monosodium salt (WST-1).
Specifications
| CYB5R2 | |
| Polyclonal | |
| Unconjugated | |
| CYB5R2 | |
| b5R.2; b5R2; CYB5R2; cytochrome b5 reductase 2; cytochrome b5 reductase b5R.2; D630003K02Rik; NADH-cytochrome b5 reductase 2; RGD1308421 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 320635, 51700 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q3KNK3, Q6BCY4 | |
| CYB5R2 | |
| Recombinant Human NADH-cytochrome b5 reductase 2 protein (1-237AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction