missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYB5R1 Antibody (OTI4B7), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71943APC
This item is not returnable.
View return policy
Description
CYB5R1 Monoclonal antibody specifically detects CYB5R1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| CYB5R1 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI4B7 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| b5R.1, B5R1, CYB5BR1, cytochrome b5 reductase 1, EC 1.6.2.2, Humb5R2, NAD(P)H:quinone oxidoreductase type 3 polypeptide A2, NQO3A2polypeptide A2 | |
| Full length human recombinant protein of human CYB5R1(NP_057327) produced in HEK293T cell. | |
| 0.1 mL | |
| metabolism | |
| 51706 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction